6-Chloro-N-methyl-2-pyridinamine
Supplier: Matrix Scientific
Synonyms:
6-Chloro-N-methylpyridin-2-amine
052705-2.5G
052705-500MG
101900-706EA
61.54
USD
101900-706
101900-708
6-Chloro-N-methyl-2-pyridinamine
6-Chloro-N-methyl-2-pyridinamine
Hazard: Irritant
Formula:
C₆H₇ClN₂ MW: 142.59 g/mol |
MDL Number:
MFCD00234196 CAS Number: 89026-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...