N2-Methyl-2,3-pyridinediamine
Supplier: Matrix Scientific
Synonyms:
N2-Methylpyridine-2,3-diamine
052701-2.5G
052701-500MG
101900-690EA
461.54
USD
101900-690
101900-692
N2-Methyl-2,3-pyridinediamine
N2-Methyl-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₆H₉N₃ MW: 123.16 g/mol |
MDL Number:
MFCD08692132 CAS Number: 5028-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...