2-[4-(2-Amino-5-fluorobenzyl)-1-piperazinyl]-1-ethanol
Supplier: Matrix Scientific
052689-2.5G
052689-500MG
101900-644EA
838.46
USD
101900-644
101900-642
2-[4-(2-Amino-5-fluorobenzyl)-1-piperazinyl]-1-ethanol
2-[4-(2-Amino-5-fluorobenzyl)-1-piperazinyl]-1-ethanol
Hazard: Irritant
Formula:
C₁₃H₂₀FN₃O |
MDL Number:
MFCD12437775 CAS Number: 1153088-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...