4-Fluoro-2-[(4-methyl-1-piperazinyl)methyl]aniline

Supplier: Matrix Scientific

052679-2.5G 052679-500MG
101900-602EA 364.62 USD
101900-602 101900-604
4-Fluoro-2-[(4-methyl-1-piperazinyl)methyl]aniline
4-Fluoro-2-[(4-methyl-1-piperazinyl)methyl]aniline

Hazard: Irritant

Formula: C₁₂H₁₈FN₃
MDL Number: MFCD12171376
CAS Number: 1153197-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR