2-Chloro-5-(2,3-dihydro-1H-indol-1-ylsulfonyl)aniline
Supplier: Matrix Scientific
052662-2.5G
052662-500MG
101900-536EA
838.46
USD
101900-536
101900-534
2-Chloro-5-(2,3-dihydro-1H-indol-1-ylsulfonyl)aniline
2-Chloro-5-(2,3-dihydro-1H-indol-1-ylsulfonyl)aniline
Hazard: Irritant
Formula:
C₁₄H₁₃ClN₂O₂S MW: 308.79 g/mol |
MDL Number:
MFCD12440248 CAS Number: 847171-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...