2-Chloro-5-[(2-ethyl-1-piperidinyl)sulfonyl]aniline

Supplier: Matrix Scientific

052657-2.5G 052657-500MG
101900-514EA 364.62 USD
101900-514 101900-516
2-Chloro-5-[(2-ethyl-1-piperidinyl)sulfonyl]aniline
2-Chloro-5-[(2-ethyl-1-piperidinyl)sulfonyl]aniline

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂S
MW: 302.83 g/mol
MDL Number: MFCD12659600
CAS Number: 1095019-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR