5-(1-Azepanylsulfonyl)-2-chloroaniline

Supplier: Matrix Scientific

052652-2.5G 052652-500MG
101900-496EA 838.46 USD
101900-496 101900-494
5-(1-Azepanylsulfonyl)-2-chloroaniline
5-(1-Azepanylsulfonyl)-2-chloroaniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂S
MDL Number: MFCD02704398
CAS Number: 326916-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR