1-[(3-Amino-4-chlorophenyl)sulfonyl]-3-pyrrolidinol

Supplier: Matrix Scientific

052643-500MG 052643-2.5G
101900-458EA 364.62 USD
101900-458 101900-460
1-[(3-Amino-4-chlorophenyl)sulfonyl]-3-pyrrolidinol
1-[(3-Amino-4-chlorophenyl)sulfonyl]-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₃S
MDL Number: MFCD13562329
CAS Number: 1220018-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR