N'-Hydroxy-2-[methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]benzenecarboximidamide
Supplier: Matrix Scientific
052637-2.5G
052637-500MG
101900-436EA
838.46
USD
101900-436
101900-434
N'-Hydroxy-2-[methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]benzenecarboximidamide
N'-Hydroxy-2-[methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]benzenecarboximidamide
Hazard: Irritant
Formula:
C₁₄H₂₁N₃O₂ |
MDL Number:
MFCD13562328 CAS Number: 1220040-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...