2-(2-Ethyl-1-piperidinyl)-N'-hydroxybenzenecarboximidamide
Supplier: Matrix Scientific
052629-2.5G
052629-500MG
101900-404EA
838.46
USD
101900-404
101900-402
2-(2-Ethyl-1-piperidinyl)-N'-hydroxybenzenecarboximidamide
2-(2-Ethyl-1-piperidinyl)-N'-hydroxybenzenecarboximidamide
Hazard: Irritant
Formula:
C₁₄H₂₁N₃O |
MDL Number:
MFCD11527410 CAS Number: 1094863-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...