2-(1-Azepanyl)-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052624-500MG 052624-2.5G
101900-382EA 364.62 USD
101900-382 101900-384
2-(1-Azepanyl)-N'-hydroxybenzenecarboximidamide
2-(1-Azepanyl)-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₃H₁₉N₃O
MDL Number: MFCD11155643
CAS Number: 1021244-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR