N'-Hydroxy-2-(4-methyl-1-piperidinyl)benzenecarboximidamide
Supplier: Matrix Scientific
052621-2.5G
052621-500MG
101900-370EA
364.62
USD
101900-370
101900-372
N'-Hydroxy-2-(4-methyl-1-piperidinyl)benzenecarboximidamide
N'-Hydroxy-2-(4-methyl-1-piperidinyl)benzenecarboximidamide
Hazard: Irritant
Formula:
C₁₃H₁₉N₃O MW: 233.32 g/mol |
MDL Number:
MFCD11155623 CAS Number: 1021243-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...