2-(Diethylamino)-N'-hydroxybenzenecarboximidamide
Supplier: Matrix Scientific
052611-2.5G
052611-500MG
101900-330EA
364.62
USD
101900-330
101900-332
2-(Diethylamino)-N'-hydroxybenzenecarboximidamide
2-(Diethylamino)-N'-hydroxybenzenecarboximidamide
Hazard: Irritant
Formula:
C₁₁H₁₇N₃O |
MDL Number:
MFCD11155630 CAS Number: 1021244-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...