N-[2-(Aminomethyl)phenyl]-N-benzyl-N-methylamine

Supplier: Matrix Scientific

052601-500MG 052601-2.5G
101900-290EA 364.62 USD
101900-290 101900-292
N-[2-(Aminomethyl)phenyl]-N-benzyl-N-methylamine
N-[2-(Aminomethyl)phenyl]-N-benzyl-N-methylamine

Hazard: Irritant

Formula: C₁₅H₁₈N₂
MDL Number: MFCD07365243
CAS Number: 893752-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR