1-[2-(Aminomethyl)phenyl]-4-piperidinol
Supplier: Matrix Scientific
052597-2.5G
052597-500MG
101900-276EA
838.46
USD
101900-276
101900-274
1-[2-(Aminomethyl)phenyl]-4-piperidinol
1-[2-(Aminomethyl)phenyl]-4-piperidinol
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O |
MDL Number:
MFCD06656912 CAS Number: 887580-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...