2-(Aminomethyl)-N,N-diethylaniline

Supplier: Matrix Scientific

052584-500MG 052584-2.5G
101900-224EA 838.46 USD
101900-224 101900-222
2-(Aminomethyl)-N,N-diethylaniline
2-(Aminomethyl)-N,N-diethylaniline

Hazard: Irritant

Formula: C₁₁H₁₈N₂
MDL Number: MFCD11155581
CAS Number: 334009-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR