3,4-Dihydro-1(2H)-quinolinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Supplier: Matrix Scientific

052581-500MG 052581-2.5G
101900-210EA 364.62 USD
101900-210 101900-212
3,4-Dihydro-1(2H)-quinolinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl
3,4-Dihydro-1(2H)-quinolinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Hazard: Irritant

Formula: C₁₆H₁₉ClN₄O
MDL Number: MFCD13562315
CAS Number: 1220033-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR