N,N-Diallyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Supplier: Matrix Scientific
052564-2.5G
052564-500MG
101900-144EA
838.46
USD
101900-144
101900-142
N,N-Diallyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N,N-Diallyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₄O MW: 282.78 g/mol |
MDL Number:
MFCD13562298 CAS Number: 1220033-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...