1-Piperidinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
Supplier: Matrix Scientific
052559-500MG
052559-2.5G
101900-122EA
364.62
USD
101900-122
101900-124
1-Piperidinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
1-Piperidinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₉ClN₄O |
MDL Number:
MFCD13562293 CAS Number: 1220039-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...