5-(2-Ethyl-1-piperidinyl)-2-(ethylsulfonyl)phenylamine
Supplier: Matrix Scientific
052546-2.5G
052546-500MG
101900-070EA
364.62
USD
101900-070
101900-072
5-(2-Ethyl-1-piperidinyl)-2-(ethylsulfonyl)phenylamine
5-(2-Ethyl-1-piperidinyl)-2-(ethylsulfonyl)phenylamine
Hazard: Irritant
Formula:
C₁₅H₂₄N₂O₂S MW: 296.43 g/mol |
MDL Number:
MFCD13562280 CAS Number: 1220029-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...