2-(Ethylsulfonyl)-5-(3-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

052537-500MG 052537-2.5G
101900-034EA 364.62 USD
101900-034 101900-036
2-(Ethylsulfonyl)-5-(3-methyl-1-piperidinyl)aniline
2-(Ethylsulfonyl)-5-(3-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₂S
MW: 282.41 g/mol
MDL Number: MFCD13562271
CAS Number: 1220033-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR