5-(2-Methyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine
Supplier: Matrix Scientific
052510-2.5G
052510-500MG
101899-926EA
364.62
USD
101899-926
101899-928
5-(2-Methyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine
5-(2-Methyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine
Hazard: Irritant
Formula:
C₁₃H₂₀N₂O₂S MW: 268.38 g/mol |
MDL Number:
MFCD13562244 CAS Number: 1220021-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...