N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052502-500MG
052502-2.5G
101899-896EA
838.46
USD
101899-896
101899-894
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinecarboxamide hydrochloride
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₅ClN₂O₂ |
MDL Number:
MFCD13562236 CAS Number: 1220039-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...