(2-Ethyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052494-500MG 052494-2.5G
101899-864EA 838.46 USD
101899-864 101899-862
(2-Ethyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
(2-Ethyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O
MDL Number: MFCD13562231
CAS Number: 1220034-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR