(2-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052485-2.5G
052485-500MG
101899-828EA
838.46
USD
101899-828
101899-826
(2-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
(2-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₃ClN₂O |
MDL Number:
MFCD13562225 CAS Number: 1220031-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...