N,N-Diethyl-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052449-2.5G
052449-500MG
101899-684EA
956.28
USD
101899-684
101899-682
N,N-Diethyl-3-piperidinecarboxamide hydrochloride
N,N-Diethyl-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O |
MDL Number:
MFCD11615736 CAS Number: 859996-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...