N-Cyclohexyl-N-methyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052437-500MG 052437-2.5G
101899-636EA 838.46 USD
101899-636 101899-634
N-Cyclohexyl-N-methyl-2-piperidinecarboxamide hydrochloride
N-Cyclohexyl-N-methyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O
MDL Number: MFCD13562189
CAS Number: 1236262-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR