N-(2-Hydroxyethyl)-N-methyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052421-500MG 052421-2.5G
101899-570EA 364.62 USD
101899-570 101899-572
N-(2-Hydroxyethyl)-N-methyl-2-piperidinecarboxamide hydrochloride
N-(2-Hydroxyethyl)-N-methyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13562176
CAS Number: 1236254-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR