(3-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride

052400-2.5G
101899-488EA 838.46 USD
101899-488
(3-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride
(3-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O
MDL Number: MFCD13562157
CAS Number: 1246172-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR