N-Butyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052396-500MG 052396-2.5G
101899-470EA 364.62 USD
101899-470 101899-472
N-Butyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride
N-Butyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562153
CAS Number: 1236263-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR