N,N-Diallyl-2-amino-3-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
052369-500MG
052369-2.5G
101899-364EA
838.46
USD
101899-364
101899-362
N,N-Diallyl-2-amino-3-phenylpropanamide hydrochloride
N,N-Diallyl-2-amino-3-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₁ClN₂O |
MDL Number:
MFCD13562128 CAS Number: 1246172-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...