2-Amino-N-(2-hydroxyethyl)-N-methyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052363-500MG 052363-2.5G
101899-338EA 364.62 USD
101899-338 101899-340
2-Amino-N-(2-hydroxyethyl)-N-methyl-3-phenylpropanamide hydrochloride
2-Amino-N-(2-hydroxyethyl)-N-methyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O₂
MDL Number: MFCD13562123
CAS Number: 1246172-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR