2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-2-methyl-1-propanone hydrochloride
Supplier: Matrix Scientific
052355-2.5G
052355-500MG
101899-308EA
838.46
USD
101899-308
101899-306
2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-2-methyl-1-propanone hydrochloride
2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-2-methyl-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O |
MDL Number:
MFCD13562115 CAS Number: 1220033-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...