2-Amino-2-methyl-N,N-dipropylpropanamide hydrochloride
Supplier: Matrix Scientific
052349-2.5G
052349-500MG
101899-284EA
838.46
USD
101899-284
101899-282
2-Amino-2-methyl-N,N-dipropylpropanamide hydrochloride
2-Amino-2-methyl-N,N-dipropylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₃ClN₂O |
MDL Number:
MFCD13562109 CAS Number: 1219971-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...