4-(2-Amino-2-methylpropanoyl)-2-piperazinone hydrochloride
Supplier: Matrix Scientific
052347-2.5G
052347-500MG
101899-276EA
838.46
USD
101899-276
101899-274
4-(2-Amino-2-methylpropanoyl)-2-piperazinone hydrochloride
4-(2-Amino-2-methylpropanoyl)-2-piperazinone hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClN₃O₂ |
MDL Number:
MFCD13562107 CAS Number: 1220034-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...