2-Amino-2-methyl-1-(3-methyl-1-piperidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052342-2.5G 052342-500MG
101899-256EA 838.46 USD
101899-256 101899-254
2-Amino-2-methyl-1-(3-methyl-1-piperidinyl)-1-propanone hydrochloride
2-Amino-2-methyl-1-(3-methyl-1-piperidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562102
CAS Number: 1220034-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR