2-Amino-N-(2-hydroxyethyl)-N-2-dimethylpropanamide hydrochloride
Supplier: Matrix Scientific
052334-500MG
052334-2.5G
101899-224EA
838.46
USD
101899-224
101899-222
2-Amino-N-(2-hydroxyethyl)-N-2-dimethylpropanamide hydrochloride
2-Amino-N-(2-hydroxyethyl)-N-2-dimethylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O₂ |
MDL Number:
MFCD13562094 CAS Number: 1220036-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...