2-Amino-N,N-2-trimethylpropanamide hydrochloride
Supplier: Matrix Scientific
052331-500MG
052331-2.5G
101899-212EA
1061.78
USD
101899-212
101899-210
2-Amino-N,N-2-trimethylpropanamide hydrochloride
2-Amino-N,N-2-trimethylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₅ClN₂O |
MDL Number:
MFCD13562091 CAS Number: 1005189-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...