2-Amino-N-benzyl-N-ethyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052329-2.5G 052329-500MG
101899-204EA 838.46 USD
101899-204 101899-202
2-Amino-N-benzyl-N-ethyl-3-methylbutanamide hydrochloride
2-Amino-N-benzyl-N-ethyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₃ClN₂O
MDL Number: MFCD13562089
CAS Number: 1236272-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR