2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-3-methyl-1-butanone hydrochloride

Supplier: Matrix Scientific

052326-500MG 052326-2.5G
101899-192EA 838.46 USD
101899-192 101899-190
2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-3-methyl-1-butanone hydrochloride
2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-3-methyl-1-butanone hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O
MDL Number: MFCD13562086
CAS Number: 1236255-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR