2-Amino-1-(4-hydroxy-1-piperidinyl)-3-methyl-1-butanone hydrochloride

Supplier: Matrix Scientific

052319-500MG 052319-2.5G
101899-162EA 364.62 USD
101899-162 101899-164
2-Amino-1-(4-hydroxy-1-piperidinyl)-3-methyl-1-butanone hydrochloride
2-Amino-1-(4-hydroxy-1-piperidinyl)-3-methyl-1-butanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD13562079
CAS Number: 1236272-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR