2-Amino-1-(1-azepanyl)-3-methyl-1-butanone hydrochloride

Supplier: Matrix Scientific

052317-500MG 052317-2.5G
101899-156EA 887.78 USD
101899-156 101899-154
2-Amino-1-(1-azepanyl)-3-methyl-1-butanone hydrochloride
2-Amino-1-(1-azepanyl)-3-methyl-1-butanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O
MDL Number: MFCD13562077
CAS Number: 1236254-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR