2-Amino-N,N-diethyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052304-2.5G 052304-500MG
101899-104EA 838.46 USD
101899-104 101899-102
2-Amino-N,N-diethyl-3-methylbutanamide hydrochloride
2-Amino-N,N-diethyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₉H₂₁ClN₂O
MDL Number: MFCD13562064
CAS Number: 1236263-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR