3-Amino-N-benzyl-N-ethylpropanamide hydrochloride
Supplier: Matrix Scientific
052300-2.5G
052300-500MG
101899-086EA
364.62
USD
101899-086
101899-088
3-Amino-N-benzyl-N-ethylpropanamide hydrochloride
3-Amino-N-benzyl-N-ethylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₉ClN₂O |
MDL Number:
MFCD13562060 CAS Number: 1220033-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...