3-Amino-1-(1-pyrrolidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-Amino-1-(pyrrolidin-1-yl)propan-1-one hydrochloride

052275-500MG 052275-2.5G
101898-986EA 364.62 USD
101898-986 101898-988
3-Amino-1-(1-pyrrolidinyl)-1-propanone hydrochloride
3-Amino-1-(1-pyrrolidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O
MW: 178.66 g/mol
MDL Number: MFCD07366746
CAS Number: 670253-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR