N-Cyclohexyl-N-ethyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052267-500MG 052267-2.5G
101898-954EA 364.62 USD
101898-954 101898-956
N-Cyclohexyl-N-ethyl-2-(methylamino)acetamide hydrochloride
N-Cyclohexyl-N-ethyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O
MDL Number: MFCD13562033
CAS Number: 1219972-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR