1-(4-Benzyl-1-piperazinyl)-2-(methylamino)-1-ethanonehydrochloride
Supplier: Matrix Scientific
052259-2.5G
052259-500MG
101898-922EA
364.62
USD
101898-922
101898-924
1-(4-Benzyl-1-piperazinyl)-2-(methylamino)-1-ethanonehydrochloride
1-(4-Benzyl-1-piperazinyl)-2-(methylamino)-1-ethanonehydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClN₃O |
MDL Number:
MFCD07289870 CAS Number: 1049765-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...