N,N-dimethyl-2-(methylamino)acetamide
Supplier: Matrix Scientific
Synonyms:
Sar-NMe₂
052245-500MG
052245-2.5G
101898-866EA
61.54
USD
101898-866
101898-868
N,N-dimethyl-2-(methylamino)acetamide
N,N-dimethyl-2-(methylamino)acetamide
Hazard: Irritant
Formula:
C₅H₁₂N₂O MW: 116.164 g/mol |
MDL Number:
MFCD00080937 CAS Number: 1857-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...