2-Amino-1-(1-piperidinyl)-1-propanone hydrochloride
Supplier: Matrix Scientific
052220-500MG
052220-2.5G
101898-766EA
364.62
USD
101898-766
101898-768
2-Amino-1-(1-piperidinyl)-1-propanone hydrochloride
2-Amino-1-(1-piperidinyl)-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O |
MDL Number:
MFCD09028974 CAS Number: 1097653-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...