2-Amino-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)acetamide hydrochloride

Supplier: Matrix Scientific

052216-500MG 052216-2.5G
101898-750EA 364.62 USD
101898-750 101898-752
2-Amino-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)acetamide hydrochloride
2-Amino-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13561991
CAS Number: 1220036-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR